48912-U Supelco

ProClin® 300

pkg of 50 mL

DOWNLOAD MSDS (PDF)

Properties

Related Categories Analytical Reagents, Analytical/Chromatography, Biocides, Biocides for In Vitro Diagnostics (IVD) Applications, Proclin More...
Quality Level   PREMIUM
contains   Alkyl Carboxylate
composition   CMIT/MIT, 3%
packaging   pkg of 50 mL
matrix   Propylene Glycol
operating pH range   2.5 - 8.5

Description

Frequently Asked Questions

Live Chat and Frequently Asked Questions are available for this Product.

Application

Biocide preservative for use in the medical in vitro diagnostic products′ market.

For use in biological diagnostic assays and IVDs only

Linkage

For additional technical details on ProClin, including key features and mechanism of action, please visit www.sigmaaldrich.com/proclin.

Legal Information

ProClin is a registered trademark of Rohm and Haas Co.

Price and Availability

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?