• USA Home
  • 48949 - EPA TCL Volatiles Mix 1

EMAIL THIS PAGE TO A FRIEND
48949 Supelco

EPA TCL Volatiles Mix 1

2000 μg/mL each component in methanol: water (9:1), analytical standard

Purchase

Properties

Related Categories Analytical Standards, Analytical/Chromatography, E, Organic Pollutant Standards, Volatile & Semivolatile Standards More...
grade   analytical standard
CofA   certificate of analysis is enclosed in each package.
packaging   ampule of 1 mL
concentration   2000 μg/mL each component in methanol: water (9:1)
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
suitability   suitable for 8260 per US EPA
format   multi-component solution
storage temp.   2-8°C

Description

Other Notes

This product is now available as a certified reference material. Please reference CRM48949.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Price and Availability

Analyte

Description

Product #

Add to Cart

Acetone SDS    
2-Butanone SDS    
2-Hexanone SDS    
4-Methyl-2-pentanone SDS    
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
RIDADR 
UN1230 - class 3 - PG 2 - Methanol
WGK Germany 
1

Documents

Certificate of Analysis

Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

CRM48949 TCL Volatiles Mix, certified reference material, 2000 μg/mL each component in methanol: water (9:1)

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?