EMAIL THIS PAGE TO A FRIEND
48960-U Supelco

γ-BHC

200 μg/mL in isooctane, analytical standard

Synonym: γ-1,2,3,4,5,6-Hexachlorocyclohexane, γ-benzene hexachloride, Lindane solution

  • CAS Number 58-89-9

  • Empirical Formula (Hill Notation) C6H6Cl6

  • Molecular Weight 290.83

  •  EC Number 208-759-1

Purchase

Properties

Related Categories 2000/60/EC, Additional Standards, Analytical Standards, Analytical/Chromatography, Application Areas and Market Segments,
grade   analytical standard
CofA   certificate of analysis is enclosed in each package.
packaging   ampule of 10 mL
concentration   200 μg/mL in isooctane
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
format   single component solution
storage temp.   room temp

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
RIDADR 
UN1262 - class 3 - PG 2 - Octanes
WGK Germany 
2

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?