Synonym:
1,1-
CAS Number 72-55-9
Beilstein Registry Number 1913355
EC Number 208-759-1
MDL number MFCD00000837
PubChem Substance ID 24872341
| Related Categories | All alphabetically sorted, Alphabetical Index of Analytical Standards, Analytical Standards, Analytical/Chromatography, Bioactive Small Molecules, |
| grade | analytical standard |
| CofA | certificate of analysis is enclosed in each package. |
| packaging | ampule of 10 mL |
| concentration | 200 μg/mL in isooctane |
| Gene Information |
mouse ... Esr1(13982) rat ... Ar(24208) |
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?