EMAIL THIS PAGE TO A FRIEND
49021 Supelco

Diazinon

analytical standard

Synonym: Dimpylate

  • CAS Number 333-41-5

  • Empirical Formula (Hill Notation) C12H21N2O3PS

  • Molecular Weight 304.35

  •  Beilstein Registry Number 273790

  •  EC Number 206-373-8

  •  MDL number MFCD00036204

  •  PubChem Substance ID 24872540

Purchase

Properties

Related Categories 2000/60/EC, Additional Standards, Alpha Index of All Pesticide Standards, Analytical Standards, Analytical/Chromatography,
grade   analytical standard
InChI Key   FHIVAFMUCKRCQO-UHFFFAOYSA-N
form   neat
packaging   ampule of 1000 mg
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
format   neat
storage temp.   room temp

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN2810 - class 6.1 - PG 3 - SMP - EHS - RQ - Toxic, liquids, organic, n.o.s., HI: all
WGK Germany 
3
RTECS 
TF3325000
Flash Point(F) 
219.2 °F
Flash Point(C) 
104 °C

Documents

Certificate of Analysis

Protocols & Articles

Articles

GC Analysis of 'Positive List' Chemicals (Mix 1) on SLB®-5ms

From our library of Articles, Sigma-Aldrich presents GC Analysis of 'Positive List' Chemicals (Mix 1) on SLB®-5ms
Keywords: Chromatography, Gas chromatography, Herbicides, Pesticides, Purification

GC Analysis of Agricultural Pesticides (Standard) on SLB®-5ms

From our library of Articles, Sigma-Aldrich presents GC Analysis of Agricultural Pesticides (Standard) on SLB®-5ms
Keywords: Agriculture, Chromatography, Gas chromatography, Gas chromatography mass spectrometry, Herbicides, Mass spectrometry, Pesticides

GC Analysis of Agricultural Pesticides on SLB®-5ms

From our library of Articles, Sigma-Aldrich presents GC Analysis of Agricultural Pesticides on SLB®-5ms
Keywords: Chromatography, Gas chromatography, Herbicides, Metabolites, Pesticides, Purification, Size-exclusion chromatography

GC Analysis of Agriculture Pesticides in Orange (Spiked) on SLB®-5ms after QuEChERS Cleanup using Primary-Secondary Amine (PSA)

From our library of Articles, Sigma-Aldrich presents GC Analysis of Agriculture Pesticides in Orange (Spiked) on SLB®-5ms after QuEChERS Cleanup using Primary-Secondary Amine (PSA)
Keywords: Chromatography, Gas chromatography, Pesticides, Purification, Sample preparations, Solid phase extractions

GC Analysis of Multiple Pesticides on SLB®-5ms after SPME using 60 μm Carbowax® Fiber

From our library of Articles, Sigma-Aldrich presents GC Analysis of Multiple Pesticides on SLB®-5ms after SPME using 60 μm Carbowax® Fiber
Keywords: Agriculture, Chromatography, Gas chromatography, Metabolites, Pesticides, Purification, Solid phase microextractions

GC Analysis of Organophosphorus Pesticides (OP-Pest) in Cabbage, Onion, and Mushrooms on SLB-5ms after SPE using Supelclean™ ENVI-Carb™ II/PSA

From our library of Articles, Sigma-Aldrich presents GC Analysis of Organophosphorus Pesticides (OP-Pest) in Cabbage, Onion, and Mushrooms on SLB-5ms after SPE using Supelclean™ ENVI-Carb™ II/PSA
Keywords: Agriculture, Centrifugation, Chromatography, Degradations, Environmental, Gas chromatography, Nitrogen phosphorus detector, Pesticides, Purification, Sample preparations, Separation, Solid phase extractions

GC Analysis of Organophosphorus Pesticides (OP-Pest) on SLB®-5ms after SPME using 60 μm Carbowax® Fiber

From our library of Articles, Sigma-Aldrich presents GC Analysis of Organophosphorus Pesticides (OP-Pest) on SLB®-5ms after SPME using 60 μm Carbowax® Fiber
Keywords: Chromatography, Gas chromatography, Metabolites, Pesticides, Purification, Solid phase microextractions

GC Analysis of Pesticides in Wine on SLB®-5ms after SPME using 85 μm Polyacrylate Fiber

From our library of Articles, Sigma-Aldrich presents GC Analysis of Pesticides in Wine on SLB®-5ms after SPME using 85 μm Polyacrylate Fiber
Keywords: Chromatography, Gas chromatography, Metabolites, Pesticides, Purification, Solid phase microextractions

LC/MS/MS of Pesticides in Oranges on Ascentis® Express C18 after Sample Prep using Supel™ Que Z-Sep/C18 QuEChERS sorbent

From our library of Articles, Sigma-Aldrich presents LC/MS/MS of Pesticides in Oranges on Ascentis® Express C18 after Sample Prep using Supel™ Que Z-Sep/C18 QuEChERS sorbent
Keywords: Chromatography, High performance liquid chromatography, Liquid chromatography mass spectrometry, Mass spectrometry, Pesticides, Sample preparations, Solid phase extractions

OSHA Method 62: GC Analysis of Organophosphorus Pesticides (OP-Pest) on SPB®-20 after Collection/Desorption using ORBO™-49P

From our library of Articles, Sigma-Aldrich presents OSHA Method 62: GC Analysis of Organophosphorus Pesticides (OP-Pest) on SPB®-20 after Collection/Desorption using ORBO™-49P
Keywords: Air Monitoring, Chromatography, Flame photometric detector, Gas chromatography, Pesticides, Purification

US EPA Method 525.2: GC Analysis of Semivolatiles on SLB®-5ms, Fast GC Analysis

From our library of Articles, Sigma-Aldrich presents US EPA Method 525.2: GC Analysis of Semivolatiles on SLB®-5ms, Fast GC Analysis
Keywords: Chromatography, Gas chromatography, Purification

US EPA Method 525.2: GC Analysis of Semivolatiles on SLB®-5ms

From our library of Articles, Sigma-Aldrich presents US EPA Method 525.2: GC Analysis of Semivolatiles on SLB®-5ms
Keywords: Chromatography, Gas chromatography, Purification

US EPA Method 8141: GC Analysis of Organophosphorus Pesticides (OP-Pest) on Equity®-5, MS Detection

From our library of Articles, Sigma-Aldrich presents US EPA Method 8141: GC Analysis of Organophosphorus Pesticides (OP-Pest) on Equity®-5, MS Detection
Keywords: Chromatography, Gas chromatography, Pesticides, Purification, Size-exclusion chromatography

US EPA Method 8141: GC Analysis of Organophosphorus Pesticides (OP-Pest) on Equity®-5, NPD Detection

From our library of Articles, Sigma-Aldrich presents US EPA Method 8141: GC Analysis of Organophosphorus Pesticides (OP-Pest) on Equity®-5, NPD Detection
Keywords: Chromatography, Gas chromatography, Nitrogen phosphorus detector, Pesticides, Purification, Size-exclusion chromatography

US EPA Method 8141: GC Analysis of Organophosphorus Pesticides (OP-Pest) on SLB®-5ms

From our library of Articles, Sigma-Aldrich presents US EPA Method 8141: GC Analysis of Organophosphorus Pesticides (OP-Pest) on SLB®-5ms
Keywords: Chromatography, Gas chromatography, Nitrogen phosphorus detector, Pesticides, Purification

US EPA Method 8141: GC Analysis of Organophosphorus Pesticides (OP-Pest) on SPB®-608

From our library of Articles, Sigma-Aldrich presents US EPA Method 8141: GC Analysis of Organophosphorus Pesticides (OP-Pest) on SPB®-608
Keywords: Chromatography, Gas chromatography, Nitrogen phosphorus detector, Pesticides, Purification

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?