EMAIL THIS PAGE TO A FRIEND
49024 Supelco

Dieldrin

analytical standard

Synonym: 1,2,3,4,10,10-Hexachloro-1,4,4a,5,6,7,8,8a-octahydro-6,7-epoxy-1,4:5,8-dimethanonaphthalene

  • CAS Number 60-57-1

  • Empirical Formula (Hill Notation) C12H8Cl6O

  • Molecular Weight 380.91

  •  Beilstein Registry Number 91396

  •  EC Number 200-484-5

  •  MDL number MFCD00135591

  •  PubChem Substance ID 24872544

Purchase

Properties

Related Categories 2000/60/EC, Additional Standards, Alpha Index of All Pesticide Standards, Analytical Standards, Analytical/Chromatography,
grade   analytical standard
vapor density   13.2 (vs air)
InChI Key   DFBKLUNHFCTMDC-PICURKEMSA-N
form   neat
packaging   ampule of 50 mg
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
mp   143-144 °C(lit.)
format   neat
storage temp.   room temp

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
RIDADR 
UN2811 - class 6.1 - PG 1 - PIH - SMP - RQ - Toxic solids, organic, n.o.s. HI: all
WGK Germany 
3
RTECS 
IO1750000

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?