Synonym: Perchloropentacyclodecane
CAS Number 2385-85-5
Empirical Formula (Hill Notation) C10Cl12
Molecular Weight 545.54
Beilstein Registry Number 5605269
EC Number 219-196-6
MDL number MFCD00167995
PubChem Substance ID 24872577
| grade | analytical standard |
| feature | standard type calibration |
| packaging | ampule of 500 mg |
| suitability | suitable for Group 22 per MISA (Canada) |
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?