49069-U Supelco

Gallocatechin solution

analytical standard, 2000 μg/mL in methanol, ampule of 0.5 mL

DOWNLOAD MSDS (PDF)

Properties

Related Categories Analytical Standards, Analytical/Chromatography, Chemical Class, Flavonoids (Catechins), Phenols More...
grade   analytical standard
CofA   certificate of analysis is enclosed in each package.
packaging   ampule of 0.5 mL
concentration   2000 μg/mL in methanol
storage temp.   −20°C

Price and Availability

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?