EMAIL THIS PAGE TO A FRIEND
49136 Supelco

Herbicides Mix 1

100 μg/mL each component in ethyl acetate, analytical standard

Purchase

Properties

Related Categories Additional Standards, All Herbicide Standards, Analytical Standards, Analytical/Chromatography, Column & Instrument Performance Test Mixes,
grade   analytical standard
CofA   certificate of analysis is enclosed in each package.
packaging   ampule of 1 mL
concentration   100 μg/mL each component in ethyl acetate
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
format   multi-component solution
storage temp.   2-8°C

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Price and Availability

Analyte

Description

Product #

Add to Cart

Atrazine SDS    
Bromacil SDS    
Cycloate SDS    
S-Ethyl-N,N-dipropylthiocarbamate SDS    
Hexazinone SDS    
Isopropalin SDS    
Metribuzin SDS    
Molinate SDS    
Oxyfluorfen SDS    
Pebulat SDS    
Terbacil SDS    
Trifluralin SDS    
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
Supplemental Hazard Statements 
Repeated exposure may cause skin dryness or cracking.
RIDADR 
UN1173 - class 3 - PG 2 - Ethyl acetate
WGK Germany 
1
Flash Point(F) 
26.6 °F
Flash Point(C) 
-3 °C

Documents

Certificate of Analysis

Protocols & Articles
Related Products

related product

Product #

Description

Add to Cart

08168 Timestrip Plus 8 °C

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?