• USA Home
  • 49475-U - Benzo(g,h,i)perylene solution

EMAIL THIS PAGE TO A FRIEND
49475-U Supelco

Benzo(g,h,i)perylene solution

certified reference material, 100 μg/mL in methylene chloride

Synonym: Benzo[ghi]perylene solution

  • CAS Number 191-24-2

  • Empirical Formula (Hill Notation) C22H12

  • Molecular Weight 276.33

  •  Beilstein Registry Number 1913029

  •  MDL number MFCD00004135

  •  PubChem Substance ID 329757461

Purchase

Properties

Related Categories Additional Standards, All Polycyclic Aromatic Hydrocarbon (PAH) Standards, Analytical Standards, Analytical/Chromatography, Certified Reference Materials (CRMs),
grade   certified reference material
InChI Key   GYFAGKUZYNFMBN-UHFFFAOYSA-N
grade   TraceCERT®
CofA   certificate of analysis is enclosed in each package.
packaging   ampule of 1 mL
availability   available only in Italy
concentration   100 μg/mL in methylene chloride
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
format   single component solution
storage temp.   room temp

Description

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO Guide 34:2009 and ISO/IEC 17025:2005. All information regarding the use of this CRM can be found on the certificate of analysis.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Warning
Hazard statements 
Precautionary statements 
Target organs 
Blood, Central nervous system, Liver
RIDADR 
UN 1593 6.1 / PGIII
WGK Germany 
2

Documents

Certificate of Analysis

Protocols & Articles

Articles

GC Analysis / Mass Spectrum of Benzo[g,h,i]Perylene (elutes at 325 °C) on SLB®-5ms, from US EPA Method 8270

From our library of Articles, Sigma-Aldrich presents GC Analysis / Mass Spectrum of Benzo[g,h,i]Perylene (elutes at 325 °C) on SLB®-5ms, from US EPA Method 8270
Keywords: Chromatography, Gas chromatography, Purification

GC Analysis of Closely Eluting Analytes from US EPA Method 8270 on SLB®-5ms (15 m x 0.10 mm I.D., 0.10 μm)

From our library of Articles, Sigma-Aldrich presents GC Analysis of Closely Eluting Analytes from US EPA Method 8270 on SLB®-5ms (15 m x 0.10 mm I.D., 0.10 μm)
Keywords: Chromatography, Flame ionization detector, Gas chromatography, Size-exclusion chromatography

GC Analysis of Closely Eluting Analytes from US EPA Method 8270 on SLB®-5ms (30 m x 0.25 mm I.D., 0.25 μm)

From our library of Articles, Sigma-Aldrich presents GC Analysis of Closely Eluting Analytes from US EPA Method 8270 on SLB®-5ms (30 m x 0.25 mm I.D., 0.25 μm)
Keywords: Chromatography, Flame ionization detector, Gas chromatography, Purification, Size-exclusion chromatography

GC Analysis of Polynuclear Aromatic Hydrocarbons (PAHs) in Salmon on SPB®-608 (20 m x 0.18 mm I.D., 0.18 μm) after QuEChERS Cleanup using Supel™ QuE Z-Sep, Fast GC Analysis

From our library of Articles, Sigma-Aldrich presents GC Analysis of Polynuclear Aromatic Hydrocarbons (PAHs) in Salmon on SPB®-608 (20 m x 0.18 mm I.D., 0.18 μm) after QuEChERS Cleanup using Supel™ QuE Z-Sep, Fast GC Analysis
Keywords: Chromatography, Gas chromatography, Gas chromatography mass spectrometry, Mass spectrometry, Purification, Sample preparations, Solid phase extractions

GC Analysis of Polynuclear Aromatic Hydrocarbons (PAHs) in Soybean Oil on SLB®-35ms after SPE using Supelclean™ EZ-POP NP

From our library of Articles, Sigma-Aldrich presents GC Analysis of Polynuclear Aromatic Hydrocarbons (PAHs) in Soybean Oil on SLB®-35ms after SPE using Supelclean™ EZ-POP NP
Keywords: Automotive, Chromatography, Combustion, Environmental, Gas chromatography, Mass selective detector, Mass spectrometry, Sample preparations, Solid phase extractions

GC Analysis of a 17-Component Polynuclear Aromatic Hydrocarbon (PAH) Mix on SLB®-IL59

From our library of Articles, Sigma-Aldrich presents GC Analysis of a 17-Component Polynuclear Aromatic Hydrocarbon (PAH) Mix on SLB®-IL59
Keywords: Chromatography, Gas chromatography, Mass selective detector

HPLC Analysis of PAHs on SUPELCOSIL™ LC-PAH

From our library of Articles, Sigma-Aldrich presents HPLC Analysis of PAHs on SUPELCOSIL™ LC-PAH
Keywords: Chromatography, Environmental, Environmental Analysis, High performance liquid chromatography, Separation

US EPA Method 525.2: GC Analysis of Semivolatiles on SLB®-5ms, Fast GC Analysis

From our library of Articles, Sigma-Aldrich presents US EPA Method 525.2: GC Analysis of Semivolatiles on SLB®-5ms, Fast GC Analysis
Keywords: Chromatography, Gas chromatography, Purification

US EPA Method 610: GC Analysis of Polynuclear Aromatic Hydrocarbons (PAHs) on SLB®-5ms

From our library of Articles, Sigma-Aldrich presents US EPA Method 610: GC Analysis of Polynuclear Aromatic Hydrocarbons (PAHs) on SLB®-5ms
Keywords: Chromatography, Gas chromatography, High performance liquid chromatography, Purification, Separation

US EPA Method 610: GC Analysis of Polynuclear Aromatic Hydrocarbons (PAHs) on SPB®-608

From our library of Articles, Sigma-Aldrich presents US EPA Method 610: GC Analysis of Polynuclear Aromatic Hydrocarbons (PAHs) on SPB®-608
Keywords: Chromatography, Gas chromatography, High performance liquid chromatography, Purification, Separation

US EPA Method 8100: GC Analysis of Polynuclear Aromatic Hydrocarbons (PAHs) on SLB®-5ms

From our library of Articles, Sigma-Aldrich presents US EPA Method 8100: GC Analysis of Polynuclear Aromatic Hydrocarbons (PAHs) on SLB®-5ms
Keywords: Chromatography, Gas chromatography, Purification

US EPA Method 8100: GC Analysis of Polynuclear Aromatic Hydrocarbons (PAHs) on SPB®-608

From our library of Articles, Sigma-Aldrich presents US EPA Method 8100: GC Analysis of Polynuclear Aromatic Hydrocarbons (PAHs) on SPB®-608
Keywords: Chromatography, Gas chromatography, Purification

US EPA Method 8270 (PAH only): GC Analysis of PAHs on SLB®-5ms (20 m x 0.18 mm I.D., 0.18 μm), Fast GC Analysis

From our library of Articles, Sigma-Aldrich presents US EPA Method 8270 (PAH only): GC Analysis of PAHs on SLB®-5ms (20 m x 0.18 mm I.D., 0.18 μm), Fast GC Analysis
Keywords: Chromatography, Gas chromatography, Purification

US EPA Method 8270 (PAH only): GC Analysis of PAHs on SLB®-5ms (20 m x 0.18 mm I.D., 0.36 μm), Fast GC Analysis

From our library of Articles, Sigma-Aldrich presents US EPA Method 8270 (PAH only): GC Analysis of PAHs on SLB®-5ms (20 m x 0.18 mm I.D., 0.36 μm), Fast GC Analysis
Keywords: Chromatography, Gas chromatography, Purification

US EPA Method 8270 (PAH only): GC Analysis of PAHs on SLB®-5ms (30 m x 0.25 mm I.D., 0.50 μm)

From our library of Articles, Sigma-Aldrich presents US EPA Method 8270 (PAH only): GC Analysis of PAHs on SLB®-5ms (30 m x 0.25 mm I.D., 0.50 μm)
Keywords: Chromatography, Gas chromatography, Purification

US EPA Method 8270 (PAH only): GC Analysis of PAHs on SLB®-5ms

From our library of Articles, Sigma-Aldrich presents US EPA Method 8270 (PAH only): GC Analysis of PAHs on SLB®-5ms
Keywords: Chromatography, Flame ionization detector, Gas chromatography, Purification

US EPA Method 8270: GC Analysis of Semivolatiles on SLB®-5ms (20 m x 0.18 mm I.D., 0.36 μm), 28 °C/min Oven Ramp, Fast GC Analysis

From our library of Articles, Sigma-Aldrich presents US EPA Method 8270: GC Analysis of Semivolatiles on SLB®-5ms (20 m x 0.18 mm I.D., 0.36 μm), 28 °C/min Oven Ramp, Fast GC Analysis
Keywords: Chromatography, Gas chromatography, Purification

US EPA Method 8270: GC Analysis of Semivolatiles on SLB®-5ms, 350 °C Final Oven Temp.

From our library of Articles, Sigma-Aldrich presents US EPA Method 8270: GC Analysis of Semivolatiles on SLB®-5ms, 350 °C Final Oven Temp.
Keywords: Chromatography, Gas chromatography

US EPA Method TO-13: GC Analysis of PAHs on SLB®-5ms

From our library of Articles, Sigma-Aldrich presents US EPA Method TO-13: GC Analysis of PAHs on SLB®-5ms
Keywords: Chromatography, Gas chromatography, Purification

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?