• USA Home
  • 4S8516 - 2-Chloroethyl vinyl ether

EMAIL THIS PAGE TO A FRIEND
4S8516 Supelco

2-Chloroethyl vinyl ether

analytical standard

  • CAS Number 110-75-8

  • Linear Formula ClCH2CH2OCH=CH2

  • Molecular Weight 106.55

  •  Beilstein Registry Number 773787

  •  EC Number 203-799-6

  •  MDL number MFCD00000973

Purchase

Properties

Related Categories A-C, Additional Standards, Alpha Sort, Analytical Standards, Analytical/Chromatography,
grade   analytical standard
vapor pressure   760 mmHg ( 109 °C)
description   Separate Source
InChI Key   DNJRKFKAFWSXSE-UHFFFAOYSA-N
form   neat
packaging   pkg of 1 × 100 mg (4S8516)
  pkg of 1 × 5 g (48516)
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
refractive index   n20/D 1.438(lit.)
bp   109 °C(lit.)
density   1.048 g/mL at 25 °C(lit.)
format   neat
storage temp.   2-8°C

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 1992 6.1(3) / PGII
WGK Germany 
3
RTECS 
KN6300000
Flash Point(F) 
60.8 °F
Flash Point(C) 
16 °C
Protocols & Articles
Related Products

related product

Product #

Description

Add to Cart

08168 Timestrip Plus 8 °C

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?