EMAIL THIS PAGE TO A FRIEND
4S8797 Supelco

EPA VOC Mix 5

analytical standard, 2000 μg/mL each component in methanol

Purchase

Properties

Related Categories 500 Series Drinking Water Methods, 8000 Series Solidwaste Methods, Additional Standards, Alpha Sort, Analytical Standards,
grade   analytical standard
description   Separate Source
CofA   certificate of analysis is enclosed in each package.
feature   standard type calibration
packaging   ampule of 1 × 1 mL
concentration   2000 μg/mL each component in methanol
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
suitability   suitable for 502.1 per US EPA
  suitable for 502.2 per US EPA
  suitable for 524.1 per US EPA
  suitable for 524.2 per US EPA
  suitable for 8010 per US EPA
  suitable for 8021 (1987) per US EPA
  suitable for 8021A (1990) per US EPA
  suitable for 8260 per US EPA
  suitable for Group 16 per MISA (Canada)
format   multi-component solution
storage temp.   2-8°C

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Price and Availability

Analyte

Description

Product #

Add to Cart

Bromodichloromethane SDS    
Dibromochloromethane SDS    
1,1-Dichloroethylene SDS    
cis-1,2-Dichloroethylene SDS    
trans-1,2-Dichloroethylene SDS    
Dichloromethane SDS    
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN1230 - class 3 - PG 2 - Methanol
WGK Germany 
2

Documents

Certificate of Analysis

Protocols & Articles
Related Products

related product

Product #

Description

Add to Cart

08168 Timestrip Plus 8 °C

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?