EMAIL THIS PAGE TO A FRIEND
4S8859 Supelco

EPA Phenols Mixture

analytical standard

Purchase

Properties

Related Categories 600 Series Wastewater Methods, Additional Standards, All Fungicides, All Herbicide Standards, Analytical Standards,
grade   analytical standard
description   Separate Source
CofA   certificate of analysis is enclosed in each package.
feature   standard type calibration
packaging   ampule of 1 × 1 mL (4S8859)
concentration   in methanol (varied)
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
suitability   suitable for 604 per US EPA
  suitable for 625 per US EPA
format   multi-component solution
storage temp.   room temp

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Price and Availability

Analyte

Description

Product #

Add to Cart

4-Chloro-3-methylphenol 2500 μg/mL SDS    
2-Chlorophenol 500 μg/mL SDS    
2,4-Dichlorophenol 500 μg/mL SDS    
2,4-Dimethylphenol 500 μg/mL SDS    
2,4-Dinitrophenol 1500 μg/mL SDS    
2-Methyl-4,6-dinitrophenol 2500 μg/mL SDS    
2-Nitrophenol 500 μg/mL SDS    
4-Nitrophenol 2500 μg/mL SDS    
Pentachlorophenol 2500 μg/mL SDS    
Phenol 500 μg/mL SDS    
2,4,6-Trichlorophenol 1500 μg/mL SDS    
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN1230 - class 3 - PG 2 - Methanol
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?