• USA Home
  • 4S8904 - EPA SS TCL Phenols Mix

EMAIL THIS PAGE TO A FRIEND
4S8904 Supelco

EPA SS TCL Phenols Mix

certified reference material, 2000 μg/mL each component in dichloromethane

Synonym: EPA TCL Phenols Mix

Purchase

Properties

Related Categories Additional Standards, Analytical Standards, Analytical/Chromatography, Certified Reference Materials (CRMs), Chromatography,
grade   certified reference material
description   Separate Source
CofA   certificate of analysis is enclosed in each package.
feature   standard type semivolatile
packaging   ampule of 1 × 1 mL (48904)
  ampule of 1 × 1 mL (4S8904)
concentration   2000 μg/mL each component in dichloromethane
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
format   multi-component solution
storage temp.   2-8°C

Description

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO Guide 34:2009 and ISO/IEC 17025:2005. All information regarding the use of this CRM can be found on the certificate of analysis.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Price and Availability

Analyte

Description

Product #

Add to Cart

4-Chloro-3-methylphenol SDS    
2-Chlorophenol SDS    
2,4-Dichlorophenol SDS    
2,4-Dimethylphenol SDS    
2,4-Dinitrophenol SDS    
2-Methyl-4,6-dinitrophenol SDS    
2-Nitrophenol SDS    
4-Nitrophenol SDS    
Pentachlorophenol SDS    
Phenol SDS    
2,4,6-Trichlorophenol SDS    
Safety & Documentation

Safety Information

Symbol 
Signal word 
Warning
Precautionary statements 
RIDADR 
UN 1593 6.1 / PGIII
WGK Germany 
2

Documents

Certificate of Analysis

Protocols & Articles
Related Products

related product

Product #

Description

Add to Cart

08168 Timestrip Plus 8 °C

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?