• USA Home
  • 4S8906 - EPA TCL Benzidines Mix

EMAIL THIS PAGE TO A FRIEND
4S8906 Supelco

EPA TCL Benzidines Mix

analytical standard, 2000 μg/mL each component in methanol

Purchase

Properties

grade   analytical standard
description   Separate Source
CofA   certificate of analysis is enclosed in each package.
feature   standard type calibration
packaging   ampule of 1 × 1 mL
concentration   2000 μg/mL each component in methanol
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
suitability   suitable for 605 per US EPA
  suitable for 625 per US EPA
  suitable for Mar ′90, Dec ′90, Aug ′91 Statement of Work per US EPA
  suitable for TCL
format   multi-component solution
storage temp.   2-8°C

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Price and Availability

Analyte

Description

Product #

Add to Cart

Benzidine SDS    
3,3′-Dichlorobenzidine SDS    
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
RIDADR 
UN1230 - class 3 - PG 2 - Methanol, solution
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

related product

Product #

Description

Add to Cart

08168 Timestrip Plus 8 °C

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?