• USA Home
  • 500496 - Low Volume Static Mixer (50-250 μL)
EMAIL THIS PAGE TO A FRIEND
500496 Supelco
Low Volume Static Mixer (50-250 μL)
housing, In-Line, PEEK
Synonym: ASI PEEK HOUSING, IN-LINE
Purchase
Properties
Related Categories ASI Static Mixers, Analytical/Chromatography, HPLC, HPLC Accessories, Low Volume Static Mixer (50-250 μL) More...
material   PEEK
fitting   in-line housing 10-32 ASI for tubing 1/16 in. O.D.
compatibility   for use with ASI low volume mixer cartridge
Description
Recommended products
Discover LiChropur reagents ideal for HPLC or LC-MS analysis
Price and Availability
All labs need water
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Content
Supelco HPLC Calculator for Isocratic or Gradient Method Transfer
Calculates conditions for transfer of an isocratic or gradient method from one HPLC column to another. Allows method scaling from microbore through preparative column range. Calculates time and solve...
Keywords: High performance liquid chromatography
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?