• USA Home
  • 500828 - Eppendorf® CH-430 column heater
EMAIL THIS PAGE TO A FRIEND
500828 Supelco
Eppendorf® CH-430 column heater
AC input 240 V
Purchase
Description
Features and Benefits
The model CH-430 column heater is like the model CH-30, except that it is wider, to accommodate up to four 30 cm long
columns (not including end fittings). The CH-430 heater also has provisions for heating two 7000 series Rheodyne valves.
Other Notes
No CE mark
Recommended products
Discover LiChropur reagents ideal for HPLC or LC-MS analysis
Footnote
Does not have a CE mark
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
Price and Availability
All labs need water
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Content
Supelco HPLC Calculator for Isocratic or Gradient Method Transfer
Calculates conditions for transfer of an isocratic or gradient method from one HPLC column to another. Allows method scaling from microbore through preparative column range. Calculates time and solve...
Keywords: High performance liquid chromatography
Related Products
related product
Product #
Description
Add to Cart
92451 Timestrip Plus 25 °C
08168 Timestrip Plus 8 °C
06693 Timestrip Plus -20 °C
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?