• USA Home
  • 502197 - EPA 508/508.1 Pesticide Mix

EMAIL THIS PAGE TO A FRIEND
502197 Supelco

EPA 508/508.1 Pesticide Mix

certified reference material, 1000 μg/mL each component in methyl tert-butyl ether

Purchase

Properties

Related Categories 500 Series Drinking Water Methods, Additional Standards, Analytical Standards, Analytical/Chromatography, Chromatography,
grade   certified reference material
  TraceCERT®
CofA   certificate of analysis is enclosed in each package.
feature   standard type calibration
packaging   ampule of 1 mL
concentration   1000 μg/mL each component in methyl tert-butyl ether
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
suitability   suitable for 508 per US EPA
format   multi-component solution
storage temp.   2-8°C

Description

Other Notes

This certified reference material (CRM) is produced and certified in accordance with ISO Guide 34:2009 and ISO/IEC 17025:2005. All information regarding the use of this CRM can be found on the certificate of analysis.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC

Price and Availability

Analyte

Description

Product #

Add to Cart

α-Chlordane SDS    
γ-Chlordane SDS    
Chlorobenzilate SDS    
Chloroneb SDS    
Chlorothalonil SDS    
Chlorthal-dimethyl SDS    
Etridiazole SDS    
Hexachlorobenzene SDS    
cis-Permethrin SDS    
trans-Permethrin SDS    
Propachlor SDS    
Trifluralin SDS    
Safety & Documentation

Safety Information

Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 2398 3 / PGII
WGK Germany 
3
Flash Point(F) 
-18.4 °F
Flash Point(C) 
-28 °C

Documents

Certificate of Analysis

Protocols & Articles

Articles

GC Analysis of cis-Chlordane Enantiomers on Astec® CHIRALDEX® G-BP

From our library of Articles, Sigma-Aldrich presents GC Analysis of cis-Chlordane Enantiomers on Astec® CHIRALDEX® G-BP
Keywords: Chromatography, Flame ionization detector, Gas chromatography, Purification

GC Analysis of α-Hexachlorocyclohexane (α-HCH) and cis-Chlordane Enantiomers on Astec® CHIRALDEX® G-BP

From our library of Articles, Sigma-Aldrich presents GC Analysis of α-Hexachlorocyclohexane (α-HCH) and cis-Chlordane Enantiomers on Astec® CHIRALDEX® G-BP
Keywords: Chromatography, Flame ionization detector, Gas chromatography, Pesticides, Purification

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?