• USA Home
  • 502227 - WSPA RM-1 Gasoline Reference Material

EMAIL THIS PAGE TO A FRIEND
502227 Supelco

WSPA RM-1 Gasoline Reference Material

analytical standard

Purchase

Properties

Related Categories ASTM Petroleum Standards, Additional Standards, Analytical Standards, Analytical/Chromatography, Chemical Class,
grade   analytical standard
packaging   pkg of 6 × 20 mL
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
format   single component solution
storage temp.   2-8°C

Description

Frequently Asked Questions

Frequently Asked Questions are available for this Product.

General description

This multipurpose reference material was developed by Scott Specialty Gases Inc. under the sponsorship of the Western States Petroleum Association (WSPA), based on specifications for California Phase II reformulated gasoline (RFG). Consensus values were established by 14 labs, including the California Air Resources Board (CARB) and are assured by WSPA.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Price and Availability


Nylon Membrane and Net Filters

Analyte

Description

Product #

Add to Cart

Benzene .982 % (w/w) SDS    
tert-Butyl methyl ether 2.07 % (w/w) SDS    
Sulfur 30.9 ppm SDS    
Aromatics 27.7 % (w/w) SDS    
Olefins 5.42 % (w/w) SDS    
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
RIDADR 
UN1203 - class 3 - PG 2 - Gasoline
WGK Germany 
3
RTECS 
DE3550000
Flash Point(F) 
-36.4 °F
Flash Point(C) 
-38 °C

Documents

Certificate of Analysis

Protocols & Articles
Related Products

related product

Product #

Description

Add to Cart

08168 Timestrip Plus 8 °C

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?