• USA Home
  • 502251 - ASTM® D5442 C16-C44 Qualitative Retention Time Mix

EMAIL THIS PAGE TO A FRIEND
502251 Supelco

ASTM® D5442 C16-C44 Qualitative Retention Time Mix

8.3 % (w/w) each component, analytical standard

Purchase

Properties

Related Categories ASTM D5442, ASTM Petroleum Standards, ASTM Petroleum and Petrochemical, Additional Standards, Analytical Standards,
grade   analytical standard
packaging   pkg of 500 mg
concentration   8.3 % (w/w) each component
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
suitability   suitable for D5442 per ASTM
format   multi-component solution
storage temp.   room temp

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

ASTM is a registered trademark of American Society for Testing and Materials

Price and Availability


Nylon Membrane and Net Filters

Analyte

Description

Product #

Add to Cart

Docosane SDS    
Dotriacontane SDS    
Eicosane SDS    
Hexacosane SDS    
Hexadecane SDS    
Hexatriacontane SDS    
Octacosane SDS    
Octadecane SDS    
Tetracontane SDS    
Tetracosane SDS    
Tetratetracontane SDS    
Triacontane SDS    
Safety & Documentation

Safety Information

Symbol 
GHS08  GHS08
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Flash Point(F) 
>212 °F
Flash Point(C) 
>100 °C

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?