• USA Home
  • 502278 - ASTM® D5442 Quantitative Linearity Standard

EMAIL THIS PAGE TO A FRIEND
502278 Supelco

ASTM® D5442 Quantitative Linearity Standard

C16-C44, 0.01 % (w/w) each component in cyclohexane, analytical standard

Purchase

Properties

Related Categories ASTM Petroleum and Petrochemical, Additional Standards, Analytical Standards, Analytical/Chromatography, Fuels and Hydrocarbons,
grade   analytical standard
packaging   pkg of 1 mL
concentration   0.01 % (w/w) each component in cyclohexane
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
suitability   suitable for D5442 per ASTM
format   multi-component solution
storage temp.   room temp

Description

Other Notes

Accurately prepared mix used to determine response factors for n-paraffins.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

ASTM is a registered trademark of American Society for Testing and Materials

Price and Availability


Nylon Membrane and Net Filters

Analyte

Description

Product #

Add to Cart

Docosane SDS    
Dotriacontane SDS    
Eicosane SDS    
Hexacosane SDS    
Hexadecane SDS    
Hexatriacontane SDS    
Octacosane SDS    
Octadecane SDS    
Tetracontane SDS    
Tetracosane SDS    
Tetratetracontane SDS    
Triacontane SDS    
Safety & Documentation

Safety Information

Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN1145 - class 3 - PG 2 - Cyclohexane
WGK Germany 
2
Flash Point(F) 
-.4 °F
Flash Point(C) 
-18 °C

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?