• USA Home
  • 502308 - EPA 8270 Acids Matrix Spiking Solution

EMAIL THIS PAGE TO A FRIEND
502308 Supelco

EPA 8270 Acids Matrix Spiking Solution

2000 μg/mL each component in methanol, analytical standard

Purchase

Properties

Related Categories 8000 Series Solidwaste Methods, Additional Standards, Analytical Standards, Analytical/Chromatography, Chromatography,
grade   analytical standard
description   for all samples
CofA   certificate of analysis is enclosed in each package.
feature   standard type matrix spiking
packaging   ampule of 1 mL
concentration   2000 μg/mL each component in methanol
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
suitability   suitable for 8270 per US EPA
format   multi-component solution
storage temp.   2-8°C

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Price and Availability

Analyte

Description

Product #

Add to Cart

4-Chloro-3-methylphenol SDS    
2-Chlorophenol SDS    
4-Nitrophenol SDS    
Pentachlorophenol SDS    
Phenol SDS    
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
RIDADR 
UN1230 - class 3 - PG 2 - Methanol
WGK Germany 
2

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?