• USA Home
  • 502723 - Frits for Precolumn MicroFilter
EMAIL THIS PAGE TO A FRIEND
502723 Supelco
Frits for Precolumn MicroFilter
stainless steel, pore size 2.0 μm
Purchase
Properties
Related Categories Analytical/Chromatography, Frits for Precolumn MicroFilter, HPLC, HPLC Accessories, Upchurch Precolumn MicroFilter and Frits,
material   stainless steel
packaging   pkg of 10 ea
mfr. no.   , C-420X
  Upchurch Scientific, Inc
pore size   2.0 μm
compatibility   for use with Precolumn Microfilter Assembiles
Description
Recommended products
Discover LiChropur reagents ideal for HPLC or LC-MS analysis
Price and Availability
All labs need water
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Content
Supelco HPLC Calculator for Isocratic or Gradient Method Transfer
Calculates conditions for transfer of an isocratic or gradient method from one HPLC column to another. Allows method scaling from microbore through preparative column range. Calculates time and solve...
Keywords: High performance liquid chromatography
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?