• USA Home
  • 502731 - Frits for Precolumn MicroFilter

EMAIL THIS PAGE TO A FRIEND
502731 Supelco

Frits for Precolumn MicroFilter

stainless steel, pore size 0.5 μm

Purchase

Properties

Related Categories Analytical/Chromatography, Frits for Precolumn MicroFilter, HPLC, HPLC Accessories, Upchurch Precolumn MicroFilter and Frits,
material   stainless steel
packaging   pkg of 10 ea
mfr. no.   Upchurch Scientific, Inc, C-425X
frit W × D × H   0.038 in. × 0.031 in. × 0.192 in.
pore size   0.5 μm
compatibility   for use with Precolumn Microfilter Assembly

Description

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Price and Availability


All labs need water
Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis

Protocols & Articles

Related Content

Supelco HPLC Calculator for Isocratic or Gradient Method Transfer

Calculates conditions for transfer of an isocratic or gradient method from one HPLC column to another. Allows method scaling from microbore through preparative column range. Calculates time and solve...
Keywords: High performance liquid chromatography

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?