• USA Home
  • 503142 - Supelpure® HC Hydrocarbon Trap
EMAIL THIS PAGE TO A FRIEND
503142 Supelco
Supelpure® HC Hydrocarbon Trap
S-trap, fitting size 1/8 in.
Purchase
Description
General description
Activated charcoal adsorbs hydrocarbons and other contaminants from carrier gases, air, and hydrogen. Operates efficiently for approximately six months when total hydrocarbons in the incoming gas average 10 ppm.
Dimensions:
• 120 cc Trap: 111/8in. (28.2 cm) long x 13/8 in. (35 mm) diameter
• 750 cc Trap: 161/2 in. (41.9 cm) long x 25/16in. (59 mm) diameter
• S-Trap: 71/2 in. (19 cm) long x 59/16 in. (14.1 cm) wide x 1/2 in. (13 mm) diameter [total bed length = 193/4 in. (50.2 cm); the extended bed length ensures prolonged contact between the gas and the adsorbent and provides greater working capacity]
Legal Information
Supelpure is a registered trademark of Sigma-Aldrich Co. LLC
Price and Availability
Safety & Documentation
Safety Information
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?