• USA Home
  • 503673 - Regulator Wall Bracket
EMAIL THIS PAGE TO A FRIEND
503673 Supelco
Regulator Wall Bracket
Single station, CGA 590
Purchase
Properties
Related Categories Analytical/Chromatography, Gas Chromatography (GC), Gas Cylinder Accessories, Gas Purification and Gas Management, Plumbing/Regulation,
feature   one pigtail on bracket with CGA connector
  Single station
fitting   CGA 590
Description
Application
for purified air
General description
Wall-mounting a gas cylinder regulator eliminates the need to handle it during cylinder change-out, thereby reducing the chance of kinking the tubing (kinked tubing next to a fitting may result in a leak). The bracket is fabricated from grade 304 stainless steel for long life, and can be used for 2-stage or 1-stage gas cylinder regulators (order separately). An integral 30 in. flexible stainless steel pigtail allows connection to a cylinder without the risk of a leak caused from kinked tubing. The CGA connector is a hand-tight design, eliminating the need for a wrench during cylinder change-out. Choose:
• CGA 580 for helium, nitrogen, and argon
• CGA 350 for hydrogen, ethane, and 5% methane in argon
• CGA 590 for compressed air
The two station bracket has 2 pigtails that join before the regulator fitting, and allows 2 gas cylinders to feed the system, extending the time between cylinder change-out.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?