51178-U Supelco
OPTI-GUARD® 1mm Guard Column
silica (orange label), pkg of 5 ea
DOWNLOAD MSDS (PDF)
Description
Frequently Asked Questions
Live Chat and Frequently Asked Questions are available for this Product.
General description
OPTI-GUARD® sets the standard for low-impact, easy to use pre-column protection. Designed for use with analytical (4.6 mm, 3.0 mm I.D.) and narrow bore (2.1 mm, 1.0 mm) columns, the patented floating stem design automatically adjusts to any manufactures tube stop depth for a zero-dead volume connection every time. The OPTI-GUARD® 1 mm requires no special connecting hardware or tools for installation.
Legal Information
OPTI-GUARD is a registered trademark of Optimize Technologies, Inc.
Price and Availability
NEW! Ascentis® Express 5 µm
HPLC Columns
New Ascentis Express 5 µm HPLC Columns!Ascentis Express columns, based on 5 µm Fused-Core® particles, offer better performance, faster and more efficient separations, compared to columns based on 3 µm and 5 µm totally porous particles, with no backpressure concerns!
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?