EMAIL THIS PAGE TO A FRIEND
51288-U Supelco

MicroCross

for use with 1/32" O. D. tubing, pkg of 1 ea

Purchase

Properties

Related Categories Analytical/Chromatography, HPLC, HPLC Accessories, MicroCross, Upchurch PEEK Micro-Fittings,
description   includes 4, microferrules for 1/32″ O.D. tubing and 4, female nuts (5/16"-24 internal threads)
packaging   pkg of 1 ea
compatibility   for use with 1/32" O. D. tubing

Description

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Price and Availability


All labs need water
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles

Related Content

Supelco HPLC Calculator for Isocratic or Gradient Method Transfer

Calculates conditions for transfer of an isocratic or gradient method from one HPLC column to another. Allows method scaling from microbore through preparative column range. Calculates time and solve...
Keywords: High performance liquid chromatography

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?