• USA Home
  • 51298-U - Upchurch Microtight® Tubing Sleeves
EMAIL THIS PAGE TO A FRIEND
51298-U Supelco
Upchurch Microtight® Tubing Sleeves
I.D. .021 in. (535 μm), natural, pkg of 10 ea
Purchase
Properties
Related Categories Analytical/Chromatography, HPLC, HPLC Accessories, Microtight Tubing Sleeves, Upchurch PEEK Micro-Fittings,
material   natural
packaging   pkg of 10 ea
I.D.   .021 in. (535 μm)
Description
General description
Use these PEEK sleeves with all Microtight fittings, to connect capillary tubing. All are 0.025 in. O.D.; normally, choose an I.D. that is 0.002-0.003 in. larger than the O.D. of the capillary tubing. Color-coded.
Recommended products
Discover LiChropur reagents ideal for HPLC or LC-MS analysis
Legal Information
Microtight is a registered trademark of IDEX Health & Science, LLC
Price and Availability
All labs need water
Safety & Documentation
Safety Information
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Documents
Certificate of Analysis
Protocols & Articles
Related Content
Supelco HPLC Calculator for Isocratic or Gradient Method Transfer
Calculates conditions for transfer of an isocratic or gradient method from one HPLC column to another. Allows method scaling from microbore through preparative column range. Calculates time and solve...
Keywords: High performance liquid chromatography
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?