EMAIL THIS PAGE TO A FRIEND
51316-U Supelco

PEEKsil Tubing

L × O.D. × I.D. 50 cm × 1/32 in. × 25 μm (.001 in.), orange, pack of 2 ea

Purchase

Properties

Related Categories 50 cm, Analytical/Chromatography, HPLC, HPLC Accessories, PEEK Tubing, PEEKsil Tubing, PTFE Tubing,
packaging   pack of 2 ea
L × O.D. × I.D.   50 cm × 1/32 in. × 25 μm (.001 in.)
color   orange

Description

General description

PEEKsil is fused silica tubing sheathed in PEEK polymer, with an effective outer diameter. The PEEK sheating is mechanically strong and has ideal characteristics for sealing with metal or polymer fittings. The fused silica core provides a consistent and rigid fluid pathway with very tight tolerances and industry-accepted chemical properties. Like traditional fused silica tubing, PEEKsil has excellent chemical compatibility and extremely low absorption characteristics, especiall when compared to stainless steel.

Please note: Do not cut this tubing. It should be used as its precut lengths because of permanent damage caused by conventional cutters.

Legal Information

PEEKSil is a trademark of SGE Analytical Science Pty Ltd.

Price and Availability


All labs need water
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles

Related Content

Supelco HPLC Calculator for Isocratic or Gradient Method Transfer

Calculates conditions for transfer of an isocratic or gradient method from one HPLC column to another. Allows method scaling from microbore through preparative column range. Calculates time and solve...
Keywords: High performance liquid chromatography

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?