• USA Home
  • 54395-U - RheBuild® Kit for SupelPRO

EMAIL THIS PAGE TO A FRIEND
54395-U Supelco

RheBuild® Kit for SupelPRO

for use with 2-Position/6-Port

Purchase

Description

Features and Benefits

Kits include all of the tools and parts you need to repair your SupelPRO unit. The parts included are those most likely to be damaged, worn, or lost. Parts may vary, depending on valve model.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

RheBuild is a registered trademark of IDEX Health & Science, LLC

SupelPRO is a trademark of Sigma-Aldrich Co. LLC

Price and Availability


All labs need water
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles

Related Content

Supelco HPLC Calculator for Isocratic or Gradient Method Transfer

Calculates conditions for transfer of an isocratic or gradient method from one HPLC column to another. Allows method scaling from microbore through preparative column range. Calculates time and solve...
Keywords: High performance liquid chromatography

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?