| Related Categories | Analytical/Chromatography, Flash Chromatography, HiTrap & PD-10 Cartridges, Pharmacia, Prepacked LPLC Columns, |
| packaging | pkg of 5 × 5 mL |
| compatibility | mode of use strong cation exchange chromatography |
| storage temp. | 2-8°C |
HiTrap® SP and HiTrap® Q columns are prepacked, ready-to-use columns, suitable for syringe or peristaltic pump use (adapters included).
Recommended flow rate: 1 mL/min
Maximum back pressure: ~40 psi (3 bar)
Chemical stability: All commonly used aqueous buffers, 1 M sodium hydroxide, 8 M urea, 6 M guanidine, 70% ethanol.
HiTrap® Q: strong anion exhange Sepharose® media for use at pH above the pI of the biomolecule (molecule will carry a negative net charge). Choose Q if the sample components are most stable above their pI.
HiTrap® SP: strong cation exhange Sepharose® media for use at pH below the pI of the biomolecule (molecule will carry a positive net charge). Choose SP if the sample components are most stable below their pI.
HiTrap is a registered trademark of GE Healthcare
Sepharose is a registered trademark of GE Healthcare
Customers Also Viewed
Fast Flow, aqueous ethanol suspension, 45-165 μm (wet), exclusion limit ~4,000,000 Da
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?