• USA Home
  • 54839 - HiTrap® Protein A Cartridge

EMAIL THIS PAGE TO A FRIEND
54839 Supelco

HiTrap® Protein A Cartridge

part volume 5 mL

Synonym: HiTrap® Affinity Cartridge

Purchase

Properties

Related Categories Antibodies, Antibody Purification and Characterization, Protein A, G and L Resins, Supplementary Products
packaging   pkg of 1 pkg
mfr. no.   Amersham Biosciences HiTrap®, 17-0403-01
  Amersham Pharmacia Bio
part volume   5 mL
compatibility   mode of use Affinity
storage temp.   2-8°C

Description

General description

HiTrap adapters and instruction manual included.

Application

HiTrap® Protein A Cartridges are used for antibody purification and characterization, protein A, G, and L resins and in supplementary products. Hitrap Protein A columns can be used for the purification of polyclonal and monoclonal antibodies, which can be used to produce nonspecific and specific tumor markers.

Legal Information

HiTrap is a registered trademark of GE Healthcare

Price and Availability


Antibody Explorer

Explore with Confidence

Amplified Detection
Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.
Protocols & Articles

Related Content

Antibody Explorer | Buy Primary & Secondary Antibodies

Monoclonal and polyclonal primary antibodies are focused on cell biology, neurobiology and molecular biology. Secondary antibodies targeting multiple host’s IgG are conjugated to alkaline phosphatase...
Keywords: Amplification, Buffers, Cell biology, Enzyme-linked immunosorbent assay, Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Molecular biology, Phosphorylations, Purification, Western blot

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?