• USA Home
  • 55055 - Rheodyne® pH Upgrade Kit
EMAIL THIS PAGE TO A FRIEND
55055 Supelco
Rheodyne® pH Upgrade Kit
for use with Valve 7125/7126
Purchase
Properties
Related Categories Analytical/Chromatography, HPLC, HPLC Accessories, Rheodyne Injectors, Sample Loops, and Accessories, Rheodyne RheBuild Kits, Fittings, Tools,
mfr. no.   , 7125-998
  Rheodyne
operating pH   0 - 14
compatibility   for use with Valve 7125/7126
Description
General description
Enables you to use your Rheodyne injector with samples at pH 0-14. Includes Tefzel rotor seal, 2 hex keys, instructions.
Legal Information
Rheodyne is a registered trademark of IDEX Health & Science, LLC
Price and Availability
All labs need water
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Content
Supelco HPLC Calculator for Isocratic or Gradient Method Transfer
Calculates conditions for transfer of an isocratic or gradient method from one HPLC column to another. Allows method scaling from microbore through preparative column range. Calculates time and solve...
Keywords: High performance liquid chromatography
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?