• USA Home
  • 56011-U - Omnifit® Column Kit
EMAIL THIS PAGE TO A FRIEND
56011-U Supelco
Omnifit® Column Kit
bed volume 110 mL, column L × I.D. 250 mm × 25 mm
Purchase
Description
General description
Omnifit glass columns are designed to be packed with resin-based 20-200 μm LC packings, and can be operated at medium pressures.
• 10 mm I.D.: 600 psi (42 bar)
• 15 mm I.D.: 300 psi (20 bar)
• 25 mm I.D.: 150 psi (10 bar)
Each Omnifit column is made from a continuous piece of precision-bore borosilicate glass, with extra-deep threaded ends and strain-free fittings. Scalloped endcaps virtually eliminate rolling and give an improved grip when opening or closing the column. Endpieces are constructed to minimize dead volume and reduce turbulence in solvent flow. A 25 μm polyethylene frit in the endpiece distributes the sample evenly over the surface of the packing bed, reducing sample dispersion before the sample enters the column.
Column Kit: 1 glass column, 2 fixed-length endpieces with Viton O-rings, 2 × 1/4-28 UNF endcaps, 1 Fittings Kit, and 1 Frit Kit. Order adjustable endpiece (flow adapter) separately.
Fittings Kit: 2 m × 1/16in. O.D. PTFE tubing with 1/4-28 UNF fitting, 0.5 m × 1/16in. O.D. PTFE tubing with 1/4-28 UNF fitting, 2 × M6 endcaps
Frit Kit: 2 × 25 μm PTFE frits, 2 each 10 μm PTFE frits and μm PE frits.
Other Notes
For aqueous mobile phases. Other diameters available as custom orders.
Legal Information
Omnifit is a registered trademark of Diba Industries
Price and Availability
All labs need water
Safety & Documentation
Safety Information
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?