• USA Home
  • 56020-U - Omnifit® Glass Column, no hardware

EMAIL THIS PAGE TO A FRIEND
56020-U Supelco

Omnifit® Glass Column, no hardware

column L × I.D. 150 mm × 15 mm

Purchase

Description

Application

Omnifit® glass columns may be used in chromatographic sample preparation, purification and traditional/classical flash chromatography.

Legal Information

Omnifit is a registered trademark of Diba Industries

Price and Availability


All labs need water

Non-Sterile Millex Syringe Filters
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?