• USA Home
  • 56023 - Omnifit® Glass Column, no hardware

EMAIL THIS PAGE TO A FRIEND
56023 Supelco

Omnifit® Glass Column, no hardware

column L × I.D. 100 mm × 25 mm

Synonym: Omnifit® column, 100mm × 25mm ID

Purchase

Description

Application

Omnifit® glass columns may be used in chromatographic sample preparation, purification and traditional/classical flash chromatography.

Legal Information

Omnifit is a registered trademark of Diba Industries

Price and Availability


All labs need water

Non-Sterile Millex Syringe Filters
Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?