• USA Home
  • 56032 - Omnifit® Standard Bore Connector
EMAIL THIS PAGE TO A FRIEND
56032 Supelco
Omnifit® Standard Bore Connector
Three-way "T" Connector
Purchase
Description
Features and Benefits
O-rings and PTFE cone included.
Application
Omnifit® Standard Bore Connector was used to construct the flow-injection system for amperometric determination of tacrine.
General description
Omnifit® Standard Bore Connector is very versatile in nature and is used for connecting various types of hardwall tubings. The standard bore connector has a manual valve operation on each end, thus the flow can be switched on or off as required. It is primarily used for fluid-line interconnection, sampling and mixing.
Legal Information
Omnifit is a registered trademark of Diba Industries
Price and Availability
All labs need water
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Peer-Reviewed Papers
15
References
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?