| Related Categories | Analytical/Chromatography, Ascentis HPLC Columns, Ascentis RP-Amide, Ascentis RP-Amide Validation Pack, HPLC, |
| packaging | pkg of 3 ea |
| L × I.D. | 25 cm × 4.6 mm |
| particle size | 5 μm |
An Ascentis RP-Amide Validation Pack makes it easy to demonstrate method reproducibility on 3 different lots. The validation pack contains a kit with 3 columns - 1 from each of 3 lots of bonded phase. And with Ascentis RP-Amide columns, you can be assured that all three columns will meet your expectations.