• USA Home
  • 567470-U - Discovery® HS PEG Supelguard Cartridge
EMAIL THIS PAGE TO A FRIEND
567470-U Supelco
Discovery® HS PEG Supelguard Cartridge
3 μm particle size, L × I.D. 2 cm × 2.1 mm
  •  eCl@ss 32110501
Purchase
Properties
Related Categories Analytical/Chromatography, Discovery HPLC Columns, Discovery HS PEG, HPLC, HPLC Columns, by Brand for Small Molecules,
packaging   pkg of 2 ea
L × I.D.   2 cm × 2.1 mm
matrix active group   polyethyleneglycol phase
particle size   3 μm
compatibility   use to protect Discovery HS PEG
mode of chromatography   Reversed phase
Description
Recommended products
Discover LiChropur reagents ideal for HPLC or LC-MS analysis
Legal Information
Discovery is a registered trademark of Sigma-Aldrich Co. LLC
Supelguard is a trademark of Sigma-Aldrich Co. LLC
Price and Availability
All labs need water
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Content
Supelco HPLC Calculator for Isocratic or Gradient Method Transfer
Calculates conditions for transfer of an isocratic or gradient method from one HPLC column to another. Allows method scaling from microbore through preparative column range. Calculates time and solve...
Keywords: High performance liquid chromatography
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?