• USA Home
  • 56762 - Eppendorf® TC-50 temperature controller
EMAIL THIS PAGE TO A FRIEND
56762 Supelco
Eppendorf® TC-50 temperature controller
AC input 120 V
Purchase
Description
Features and Benefits
Controls a CH-30 Eppendorf heater from ambient temperature to 150 °C. Temperature is set via a 10 turn potentiometer. The set point and actual temperature are displayed on an LED display. Includes remote heater on/off capability and solid state control.
Other Notes
No CE mark
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
Price and Availability
All labs need water
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Content
Supelco HPLC Calculator for Isocratic or Gradient Method Transfer
Calculates conditions for transfer of an isocratic or gradient method from one HPLC column to another. Allows method scaling from microbore through preparative column range. Calculates time and solve...
Keywords: High performance liquid chromatography
Related Products
related product
Product #
Description
Add to Cart
92451 Timestrip Plus 25 °C
08168 Timestrip Plus 8 °C
06693 Timestrip Plus -20 °C
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?