• USA Home
  • 57093 - Visidry Long Stem Flow Control Knob
EMAIL THIS PAGE TO A FRIEND
57093 Supelco
Visidry Long Stem Flow Control Knob
pk of 6
Purchase
Description
General description
If you have equipped your Visiprep Vacuum Manifold with long stem flow control valves (57048), these control knobs will enable you to attach the Visidry Drying Attachment without removing the long stem valves.
Note: These knobs cannot be used with a 24-port manifold being used to process 12mL, 20mL, or 60mL tubes.
Legal Information
Visidry is a trademark of Sigma-Aldrich Co. LLC
Price and Availability
All labs need water
Non-Sterile Millex Syringe Filters
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Protocols & Articles
Related Content
SPE Cartridges (Tubes) - Phase Selection Guide
The SPE Tube Configuraton Guide will help you choose the appropriate bed weight and cartridge dimensions.
Keywords: Agriculture, Calorimetry, Environmental, Fractionation, High performance liquid chromatography, Liquid chromatography mass spectrometry, Mass spectrometry, Pesticides, Pharmaceutical, Solid phase extractions
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?