• USA Home
  • 57204 - Supelclean LC-SCX SPE Bulk Packing

EMAIL THIS PAGE TO A FRIEND
57204 Supelco

Supelclean LC-SCX SPE Bulk Packing

pkg of 100 g

Purchase

Properties

Related Categories Analytical/Chromatography, Ion-Exchange Supelclean SPE Products, Sample Preparation & Purification, Solid Phase Extraction (SPE), Supelclean ENVI and Supelclean SPE Cartridges,
packaging   pkg of 100 g
matrix active group   SCX phase
  SCX
mode of chromatography   Ion exchange
  Ion exchange

Description

Legal Information

Supelclean is a trademark of Sigma-Aldrich Co. LLC

Price and Availability


All labs need water

Non-Sterile Millex Syringe Filters
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
RIDADR 
UN 1759 8 / PGIII
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles

Articles

Solid Phase Extraction: Ion Exchange Methodology

In order for electrostatic retention to occur, both analyte and sorbent functional groups must be in their ionized form. This is done through strict pH control of the sample matrix. For basic analyte...
Keywords: Agriculture, Buffers, Herbicides, Ion Exchange, Pharmaceutical, Solid phase extractions

Solid Phase Extraction: Normal Phase Methodology

In order for polar retention to occur between the sorbent and the sample, the analyte must be introduced to the SPE device in a non-polar sample or mobile phase environment. Therefore, typical sample...
Keywords: Cosmetics, Fractionation, Gas chromatography, Separation, Solid phase extractions, Solvents, Sonication

Related Content

SPE Cartridges (Tubes) - Phase Selection Guide

The SPE Tube Configuraton Guide will help you choose the appropriate bed weight and cartridge dimensions.
Keywords: Agriculture, Calorimetry, Environmental, Fractionation, High performance liquid chromatography, Liquid chromatography mass spectrometry, Mass spectrometry, Pesticides, Pharmaceutical, Solid phase extractions

Solid Phase Extraction: Fundamentals and Recent Developments [Webinar]

Solid Phase Extraction (SPE) has become a popular technique for sample preparation due to its convenience, effectiveness, relatively low solvent usage and versatility. The focus of this webinar is on...
Keywords: Sample preparations, Solid phase extractions

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?