• USA Home
  • 57207 - Supelclean LC-Alumina-B SPE Bulk Packing

EMAIL THIS PAGE TO A FRIEND
57207 Supelco

Supelclean LC-Alumina-B SPE Bulk Packing

pkg of 100 g

Purchase

Properties

Related Categories Adsorption Supelclean SPE Products, Analytical/Chromatography, Sample Preparation & Purification, Solid Phase Extraction (SPE), Supelclean ENVI and Supelclean SPE Cartridges,
packaging   pkg of 100 g
matrix   LC-Alumina-B
matrix active group   Alumina phase
diameter   60-325 mesh
compatibility   for use with Basic pH (8.5)
mode of chromatography   Normal phase

Description

Legal Information

Supelclean is a trademark of Sigma-Aldrich Co. LLC

Price and Availability


Laboratory Gloves

All labs need water

Non-Sterile Millex Syringe Filters
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
nwg
RTECS 
BD1200000

Documents

Certificate of Analysis

Protocols & Articles

Related Content

SPE Cartridges (Tubes) - Phase Selection Guide

The SPE Tube Configuraton Guide will help you choose the appropriate bed weight and cartridge dimensions.
Keywords: Agriculture, Calorimetry, Environmental, Fractionation, High performance liquid chromatography, Liquid chromatography mass spectrometry, Mass spectrometry, Pesticides, Pharmaceutical, Solid phase extractions

Solid Phase Extraction: Fundamentals and Recent Developments [Webinar]

Solid Phase Extraction (SPE) has become a popular technique for sample preparation due to its convenience, effectiveness, relatively low solvent usage and versatility. The focus of this webinar is on...
Keywords: Sample preparations, Solid phase extractions

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?