• USA Home
  • 57489 - alphaBond HPLC Column

EMAIL THIS PAGE TO A FRIEND
57489 Supelco

alphaBond HPLC Column

L × I.D. 30 cm × 3.9 mm

Purchase

Properties

Related Categories Analytical/Chromatography, Established Column Lines from Other Manufacturers, HPLC, HPLC Columns, by Brand for Small Molecules, alphaBond Columns More...
mfr. no.   alphaBond C18
parameter   ≤70 °C temp. range
application(s)   HPLC: suitable
L × I.D.   30 cm × 3.9 mm
matrix active group   C18 (octadecyl) phase
particle size   10 μm
pore size   125 Å
surface area   300 m2/g
mode of chromatography   Reversed phase

Description

Legal Information

alphaBond is a trademark of Almatis

Price and Availability


All labs need water
Safety & Documentation

Safety Information

Symbol 
GHS08  GHS08
Signal word 
Danger
Hazard statements 
Precautionary statements 
WGK Germany 
nwg

Documents

Certificate of Analysis

Protocols & Articles

Related Content

Supelco HPLC Calculator for Isocratic or Gradient Method Transfer

Calculates conditions for transfer of an isocratic or gradient method from one HPLC column to another. Allows method scaling from microbore through preparative column range. Calculates time and solve...
Keywords: High performance liquid chromatography

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?