• USA Home
  • 581696-U - Ascentis Phenyl Validation Pack

EMAIL THIS PAGE TO A FRIEND
581696-U Supelco

Ascentis Phenyl Validation Pack

5 μm particle size, L × I.D. 25 cm × 4.6 mm, Validation Pack, pkg of 3 ea

  •  eCl@ss 32110501

Purchase

Properties

Related Categories Analytical/Chromatography, Ascentis HPLC Columns, Ascentis Phenyl, Ascentis Phenyl Validation Pack, HPLC,
packaging   pkg of 3 ea
application(s)   HPLC: suitable
L × I.D.   25 cm × 4.6 mm
matrix active group   phenyl phase
particle size   5 μm
pore size   100 Å
suitability   suitable for L11 per USP
mode of chromatography   Reversed phase

Description

General description

An Ascentis Phenyl Validation Pack makes it easy to demonstrate method reproducibility on 3 different lots. The validation pack contains a kit with 3 columns - 1 from each of 3 lots of bonded phase. And with Ascentis Phenyl columns, you can be assured that all three columns will meet your expectations.

Price and Availability


All labs need water
Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis

Protocols & Articles

Related Content

Supelco HPLC Calculator for Isocratic or Gradient Method Transfer

Calculates conditions for transfer of an isocratic or gradient method from one HPLC column to another. Allows method scaling from microbore through preparative column range. Calculates time and solve...
Keywords: High performance liquid chromatography

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?