58299 Supelco

LC-CN/LC-PCN Test Mix

in acetonitrile: water (varied conc.), solution, analytical standard

DOWNLOAD MSDS (PDF)

Properties

Related Categories Alphabetical Index of Analytical Standards, Analytical Standards, Analytical/Chromatography, Chromatography, Column Test Mixes (HPLC),
grade   analytical standard
form   solution
packaging   ampule of 1 mL
concentration   in acetonitrile: water (varied conc.)
compatibility   use to QA LC-CN, LC-PCN columns, any weakly hydrophobic phase

Price and Availability

Analyte

Description

Product #

Add to Cart

Acetophenone 7 μg/mL    
Benzene 750 μg/mL    
Toluene 775 μg/mL    
Uracil 7 μg/mL    

Customers Also Viewed

in methanol: water (3:2) (varied conc.), analytical standard

in acetonitrile: water (25:75), analytical standard

in methanol: water (3:2) (varied conc.), analytical standard

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?