• USA Home
  • 58458AST - CHIRALPAK® HSA HPLC Guard Column

EMAIL THIS PAGE TO A FRIEND
58458AST Supelco

CHIRALPAK® HSA HPLC Guard Column

5 μm particle size, L × I.D. 1 cm × 3 mm, pk of 2 ea

  •  eCl@ss 32110501

Purchase

Properties

Related Categories Analytical/Chromatography, CHIRALPAK HSA, Chiral HPLC Columns, HPLC, Protein-based Chiral HPLC Columns More...
packaging   pk of 2 ea
application(s)   HPLC: suitable
L × I.D.   1 cm × 3 mm
matrix active group   human serum albumin phase
particle size   5 μm
suitability   suitable for L79 per USP
mode of chromatography   Chiral

Description

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

CHIRALPAK is a registered trademark of Daicel Corp.

Price and Availability


All labs need water
Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis

Protocols & Articles

Related Content

Supelco HPLC Calculator for Isocratic or Gradient Method Transfer

Calculates conditions for transfer of an isocratic or gradient method from one HPLC column to another. Allows method scaling from microbore through preparative column range. Calculates time and solve...
Keywords: High performance liquid chromatography

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?