• USA Home
  • 59602-U - Discovery® BIO PolyMA-WAX Column

EMAIL THIS PAGE TO A FRIEND
59602-U Supelco

Discovery® BIO PolyMA-WAX Column

5 μm particle size, L × I.D. 5 cm × 4.6 mm

  •  eCl@ss 32110501

Purchase

Properties

Related Categories Analytical/Chromatography, Discovery BIO HPLC Columns, Discovery BIO Ion Exchange Columns, Discovery BIO PolyMA Ion-Exchange Columns, HPLC,
parameter   4-50 °C temp. range
  735 psi max. pressure
application(s)   HPLC: suitable
L × I.D.   5 cm × 4.6 mm
surface coverage   surface coverage 0.3 meq/g
matrix   polymethacrylate, spherical, monodispersed particle platform
matrix active group   DEAE (diethylaminoethyl) phase
particle size   5 μm
pore size   1000 Å
operating pH range   2 - 11
suitability   suitable for L23 per USP
compatibility   mode of use anion-exchange
mode of chromatography   Anion exchange

Description

Application

For use at pH greater than the protein isoelectric point (pI), usually at pH 7 or higher

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

Discovery is a registered trademark of Sigma-Aldrich Co. LLC

Price and Availability


All labs need water
Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.
Protocols & Articles

Articles

HPLC Analysis of Hemoglobin Variants on Discovery® BIO PolyMA-WAX

From our library of Articles, Sigma-Aldrich presents HPLC Analysis of Hemoglobin Variants on Discovery® BIO PolyMA-WAX
Keywords: Chromatography, High performance liquid chromatography

HPLC Analysis of Human Growth Hormone (hGH) on Discovery® BIO PolyMA-WAX

From our library of Articles, Sigma-Aldrich presents HPLC Analysis of Human Growth Hormone (hGH) on Discovery® BIO PolyMA-WAX
Keywords: Chromatography, Degradations, High performance liquid chromatography, Hormones

HPLC Analysis of Proteins on Discovery® BIO PolyMA-WAX (Analyte Set #1)

From our library of Articles, Sigma-Aldrich presents HPLC Analysis of Proteins on Discovery® BIO PolyMA-WAX (Analyte Set #1)
Keywords: Chromatography, High performance liquid chromatography, Separation

HPLC Analysis of Superoxide Dismutase on Discovery® BIO PolyMA-WAX

From our library of Articles, Sigma-Aldrich presents HPLC Analysis of Superoxide Dismutase on Discovery® BIO PolyMA-WAX
Keywords: Chromatography, High performance liquid chromatography

Related Content

Supelco HPLC Calculator for Isocratic or Gradient Method Transfer

Calculates conditions for transfer of an isocratic or gradient method from one HPLC column to another. Allows method scaling from microbore through preparative column range. Calculates time and solve...
Keywords: High performance liquid chromatography

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?