• USA Home
  • 813353 - TSKgel® OligoDNA RP HPLC Column

EMAIL THIS PAGE TO A FRIEND
813353 Supelco

TSKgel® OligoDNA RP HPLC Column

phase C18 (octadecyl), L × I.D. 15 cm × 7.8 mm, 5 μm particle size

Synonym: TSKgel® Reversed Phase HPLC Column

Purchase

Properties

Related Categories Analytical/Chromatography, HPLC, HPLC & FPLC Columns for Biopolymer Separations, TSKgel HPLC Columns, TOYOPEARL Bulk Media & Accessories, TSKgel OligoDNA RP Columns,
material   stainless steel hardware
mfr. no.   Tosoh OligoDNA RP, 13353
extent of labeling   10% Carbon, not endcapped
parameter   salt concentration
  0-100% organic solvent
  0-45 °C temperature
  120 bar max. pressure
  3.2 mL/min flow rate
L × I.D.   15 cm × 7.8 mm
matrix   silica particle platform
matrix active group   C18 (octadecyl) phase
particle size   5 μm
operating pH   2.0 - 7.5
suitability   suitable for L1 per USP
  suitable for L1
mode of chromatography   Reversed phase

Description

Physical form

Shipped in 50% acetonitrile - 50% water.

General description

Specifically designed for purification of oligonucleotides, RNA and DNA fragments (up to 500mer), TSKgel OligoDNA RP can provide excellent separations of samples with very similar sequences. The 250Å pore size provides excellent kinetics for molecules with helix shape structures processing large radii of gyration. The 5 micron particle size provides a minumum of 7,000 plates per 15 centimeter column.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

TSKgel is a registered trademark of Tosoh Corporation

Price and Availability


All labs need water
Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis

Protocols & Articles

Related Content

Supelco HPLC Calculator for Isocratic or Gradient Method Transfer

Calculates conditions for transfer of an isocratic or gradient method from one HPLC column to another. Allows method scaling from microbore through preparative column range. Calculates time and solve...
Keywords: High performance liquid chromatography

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?